missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepine, MedChemExpress

Product Code. 16482066
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482066 500 mg 500mg
16478186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482066 Supplier Medchem Express Supplier No. HYB0246
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Carbamazepine, a sodium channel blocker, is an anticonvulsant drug.

Carbamazepine, a sodium channel blocker, is an anticonvulsant drug.

Specifications

Chemical Name or Material Carbamazepine
Physical Form Powder
CAS 298-46-4
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C15H12N2O
For Use With (Application) Cancer-programmed cell death
Synonym CBZ NSC 169864
Solubility Information DMSO : 100 mg/mL (423.24 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(N1C2=CC=CC=C2C=CC3=CC=CC=C31)N
Molecular Weight (g/mol) 236.27
Formula Weight 236.27
Percent Purity 99.75%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.