missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbamazepine EP Impurity C, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Carbamazepine EP Impurity C
Specifications
Specifications
| Chemical Name or Material | (5H-Dibenzo[b,f]azepin-5-yl-carbonyl)urea (N-Carbamoylcarbamazepine) |
| CAS | 1219170-51-0 |
| Analyte or Component Names | (5H-Dibenzo[b,f]azepin-5-yl-carbonyl)urea (N-Carbamoylcarbamazepine) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H13 N3 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Carbamazepine Imp. C (EP), (5H-Dibenzo[b,f]azepin-5-yl-carbonyl)urea, N-Carbamoylcarbamazepine, Carbamazepine Impurity C |
| InChI Formula | InChI=1S/C16H13N3O2/c17-15(20)18-16(21)19-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)19/h1-10H,(H3,17,18,20,21) |
| SMILES | NC(=O)NC(=O)N1c2ccccc2C=Cc3ccccc13 |
| IUPAC Name | N-carbamoylbenzo[b][1]benzazepine-11-carboxamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction