missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Carbamazepine Monoclonal antibody specifically detects Carbamazepine in Non-species specific samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Carbamazepine |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1014505 |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | 298-46-4, Carbazepin, Epitol, Tegretol |
| Host Species | Mouse |
| Immunogen | Synthetic Hapten version of Carbamazepine conjugated to KLH. |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?