missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepine Antibody (1014505) [Alexa Fluor™ 488], Novus Biologicals™

Product Code. 30689868 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689868 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30689868 Supplier Novus Biologicals Supplier No. MAB11685AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carbamazepine Monoclonal antibody specifically detects Carbamazepine in Non-species specific samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbamazepine
Applications ELISA
Classification Monoclonal
Clone 1014505
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias 298-46-4, Carbazepin, Epitol, Tegretol
Host Species Mouse
Immunogen Synthetic Hapten version of Carbamazepine conjugated to KLH.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 0
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.