Learn More
Carbamazepine, 98%, Thermo Scientific™
£96.65 - £320.00
Chemical Identifiers
| CAS | 298-46-4 |
|---|---|
| Molecular Formula | C15H12N2O |
| Molecular Weight (g/mol) | 236.27 |
| MDL Number | MFCD00005073 |
| InChI Key | FFGPTBGBLSHEPO-UHFFFAOYSA-N |
| Synonym | carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston |
| PubChem CID | 2554 |
| ChEBI | CHEBI:3387 |
| IUPAC Name | 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide |
| SMILES | NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15434929
|
Thermo Scientific Alfa Aesar
J62590.14 |
25g |
£107.00 £96.65 / 25g Save £10.35 9% Off |
Estimated Shipment: 11-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15444929
|
Thermo Scientific Alfa Aesar
J62590.22 |
100g |
£320.00
100g |
Estimated Shipment: 25-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in ethanol, acetone, propylene glycol, chloroform, dimethyl sulfoxide, methanol, dimethyl fluoride,ethylene glycol and monoethyl ether. Slightly soluble in glacial acetic acid. Insoluble in water.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 298-46-4 | |
| 236.27 | |
| FFGPTBGBLSHEPO-UHFFFAOYSA-N | |
| 2554 | |
| 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide |
| C15H12N2O | |
| MFCD00005073 | |
| carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston | |
| CHEBI:3387 | |
| NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12 |
Specifications
| 298-46-4 | |
| MFCD00005073 | |
| carbamazepine, tegretol, 5h-dibenzo b,f azepine-5-carboxamide, carbamazepen, carbazepine, finlepsin, carbamezepine, equetro, tegretal, biston | |
| FFGPTBGBLSHEPO-UHFFFAOYSA-N | |
| 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene-2-carboxamide | |
| 2554 | |
| 236.27 | |
| Carbamazepine |
| C15H12N2O | |
| 14,1781 | |
| Soluble in ethanol,acetone,propylene glycol,chloroform,dimethyl sulfoxide,methanol,dimethyl fluoride,ethylene glycol and monoethyl ether. Slightly soluble in glacial acetic acid. Insoluble in water. | |
| NC(=O)N1C2=CC=CC=C2C=CC2=CC=CC=C12 | |
| 236.27 | |
| CHEBI:3387 | |
| 98% |
Safety and Handling
GHS H Statement
H334-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P272-P280g-P285-P301+P312-P302+P352-P304+P341-P330-P333+P313-P342+P311-P363-P501c
H302-H317-H334
missing translation for 'einecsNumber' : 206-062-7
missing translation for 'rtecsNumber' : HN8225000
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.