missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepine 10,11-Epoxide, Mikromol™

Product Code. 17932456
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932456 100 mg 100mg
17923046 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17932456 Supplier Mikromol Supplier No. MM0076.01
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Carbamazepine 10,11-Epoxide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbamazepine-10,11-epoxide
CAS 36507-30-9
Analyte or Component Names Carbamazepine-10,11-epoxide
Shipping Condition Ice pack (-20°C)
Molecular Formula C15 H12 N2 O2
Certifications/Compliance ISO 17025
Synonym Carbamazepine 10,11-Epoxide, 1a,10b-Dihydro-6H-dibenz[b,f]oxireno[d]azepine-6-carboxamide, 10,11-Dihydro-5H-dibenz[b,f]azepine-5-carboxamide 10,11-epoxide, 10,11-Epoxycarbamazepine, Carbamazepine 10,11-epoxide, Carbamazepine 10,11-oxide, GP 49-023
InChI Formula InChI=1S/C15H12N2O2/c16-15(18)17-11-7-3-1-5-9(11)13-14(19-13)10-6-2-4-8-12(10)17/h1-8,13-14H,(H2,16,18)
SMILES NC(=O)N1c2ccccc2C3OC3c4ccccc14
IUPAC Name 1a,10b-Dihydro-6H-dibenz[b,f]oxireno[d]azepine-6-carboxamide
Molecular Weight (g/mol) 252.27
Quantity 100 mg
Formula Weight 252.0899 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.