missing translation for 'onlineSavingsMsg'
Learn More

Carbamazepine 10,11-epoxide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30326498
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326498 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326498 Supplier Dr. Ehrenstorfer Supplier No. DREA10968550AL100
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbamazepine-10,11-epoxide
CAS 36507-30-9
Analyte or Component Names Carbamazepine-10,11-epoxide
Shipping Condition +20°C
Molecular Formula C15 H12 N2 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Carbamazepine 10,11-Epoxide, 1a,10b-Dihydro-6H-dibenz[b,f]oxireno[d]azepine-6-carboxamide, 10,11-Dihydro-5H-dibenz[b,f]azepine-5-carboxamide 10,11-epoxide, 10,11-Epoxycarbamazepine, Carbamazepine 10,11-epoxide, Carbamazepine 10,11-oxide, GP 49-023
InChI Formula InChI=1S/C15H12N2O2/c16-15(18)17-11-7-3-1-5-9(11)13-14(19-13)10-6-2-4-8-12(10)17/h1-8,13-14H,(H2,16,18)
SMILES NC(=O)N1c2ccccc2C3OC3c4ccccc14
IUPAC Name 1a,10b-Dihydro-6H-dibenz[b,f]oxireno[d]azepine-6-carboxamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 252.27
Quantity 1 mL
Formula Weight 252.0899 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.