missing translation for 'onlineSavingsMsg'
Learn More

Carbamate Pesticide Mixture 673 79 μg/mL (100 mg/kg) in Methanol, Dr. Ehrenstorfer

Product Code. 30313965
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30313965 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30313965 Supplier Dr. Ehrenstorfer Supplier No. DREA50000673ME
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Aminocarb∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Fenobucarb∣Indoxacarb∣Isoprocarb∣Methiocarb∣Methomyl∣Metolcarb∣Pirimicarb∣Propoxur
CAS 116-06-3, 1646-88-4, 1646-87-3, 2032-59-9, 63-25-2, 1563-66-2
Analyte or Component Names Aldicarb∣Aldicarb-sulfone∣Aldicarb-sulfoxide∣Aminocarb∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Fenobucarb∣Indoxacarb∣Isoprocarb∣Methiocarb∣Methomyl∣Metolcarb∣Pirimicarb∣Propoxur
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Aldicarb 79 μg/mL∣Aldicarb-sulfone 79 μg/mL∣Aldicarb-sulfoxide 79 μg/mL∣Aminocarb 79 μg/mL∣Carbaryl 79 μg/mL∣Carbofuran 79 μg/mL∣Carbofuran-3-hydroxy 79 μg/mL∣Fenobucarb 79 μg/mL∣Indoxacarb 79 μg/mL∣Isoprocarb 79 μg/mL∣Methiocarb 79 μg/mL∣Methomyl 79 μg/mL∣Metolcarb 79 μg/mL∣Pirimicarb 79 μg/mL∣Propoxur 79 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.