missing translation for 'onlineSavingsMsg'
Learn More

Carbadox (Technical Grade), TRC

Product Code. 30368454
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368454 1 g 1g
30352967 100 mg 100mg
30353288 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368454 Supplier Toronto Research Chemicals Supplier No. TRCC2758251G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6804-07-5
Molecular Formula C11 H10 N4 O4
Molecular Weight (g/mol) 262.22
Synonym Carbazic acid, 3-(2-quinoxalinylmethylene)-, methyl ester, N,N'-dioxide (7CI), Carbazic acid, 3-(2-quinoxalinylmethylene)-, methyl ester, N1,N4-dioxide (8CI), Hydrazinecarboxylic acid, (2-quinoxalinylmethylene)-, methyl ester, N,N'-dioxide, Hydrazinecarboxylic acid, [(1,4-dioxido-2-quinoxalinyl)methylene]-, methyl ester (9CI), 2-Formylquinoxaline-1,4-dioxide carbomethoxyhydrazone, Carbadox, Fortigro, GS 6244, Getroxel, Mecadox, Methyl 3-(2-quinoxalinylmethylene)carbazate N1,N4-dioxide, Quinoxaline-2-carbomethoxyhydrazone 1,4-dioxide
IUPAC Name methyl N-[(E)-(1,4-dioxidoquinoxaline-1,4-diium-2-yl)methylideneamino]carbamate
SMILES COC(=O)N\N=C\c1c[n+]([O-])c2ccccc2[n+]1[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H10N4O4/c1-19-11(16)13-12-6-8-7-14(17)9-4-2-3-5-10(9)15(8)18/h2-7H,1H3,(H,13,16)/b12-6+
Formula Weight 262.0702
Chemical Name or Material Carbadox
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.