missing translation for 'onlineSavingsMsg'
Learn More

Carbadox, MedChemExpress

Product Code. 16458736
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458736 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458736 Supplier Medchem Express Supplier No. HYB1340
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Carbadox is a quinoxaline-di-N-oxide antibiotic compound which is widely fed to nursery-age pigs to control enteric diseases and improve feed efficiency.

Carbadox is a quinoxaline-di-N-oxide antibiotic compound which is widely fed to nursery-age pigs to control enteric diseases and improve feed efficiency.

Specifications

Chemical Name or Material Carbadox
Physical Form Solid
CAS 6804-07-5
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H10N4O4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 3.57 mg/mL (13.61 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES [O-][N+]1=C2C=CC=CC2=[N+]([O-])C(/C=N/NC(OC)=O)=C1
Molecular Weight (g/mol) 262.22
Formula Weight 262.22
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.