missing translation for 'onlineSavingsMsg'
Learn More

Carbachol, 99%, Thermo Scientific™

Carbachol, CAS # 51-83-2, is a parasympathomimetic that acts on acetylcholine receptors.

Brand:  Thermo Scientific Alfa Aesar L06674.06

Quantity 5g

 View more versions of this product

Product Code. 15448859

Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description
• Carbachol is an ammonium salt and a choline ester that acts on both muscarinic and nicotinic receptors
• Carbachol activates the sympathetic nervous system; it leads to intracellular increases in Ca2+ and activates associated signaling pathways
• Carbachol is not degraded by the cholinesterases
Application
• Carbachol can inhibit apoptotic cell death, making it useful for the study of cell survival and growth in laboratory settings
• Carbachol can be used for laboratory research of any biological pathway that depends on acetylcholine, including sleep cycles and motor control
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

51-83-2
182.65
AIXAANGOTKPUOY-UHFFFAOYSA-N
5831
2-(trimethylazaniumyl)ethyl carbamate chloride
C6H15ClN2O2
MFCD00012011
carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
CHEBI:3385
[Cl-].C[N+](C)(C)CCOC(N)=O

Specifications

51-83-2
MFCD00012011
UN2811
Hygroscopic
carbachol, carbamoylcholine chloride, carbachol chloride, carbacholine, carbocholine, jestryl, doryl, carbamylcholine chloride, miostat, isopto carbachol
AIXAANGOTKPUOY-UHFFFAOYSA-N
2-(trimethylazaniumyl)ethyl carbamate chloride
5831
182.65
Carbachol
C6H15ClN2O2
5g
3917459
14,1779
Soluble in water and dimethyl sulfoxide.
[Cl-].C[N+](C)(C)CCOC(N)=O
182.65
CHEBI:3385
99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.