missing translation for 'onlineSavingsMsg'
Learn More

(±)-Carazolol-d7, TRC

Product Code. 30134276
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134276 25 mg 25mg
17918529 2.5 mg 2.5mg
17959399 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30134276 Supplier Toronto Research Chemicals Supplier No. TRCC17580725MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173021-02-7
Molecular Formula C18 2H7 H15 N2 O2
Molecular Weight (g/mol) 305.42
Synonym 1-(9H-Carbazol-4-yloxy)-3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)propan-2-ol, Carazolol D7, Carazolol-D7 (N-isopropyl-D7), (±)-Carazolol-d7 (iso-propyl-d7), (±)-Carazolol-d7, 1-(9H-Carbazol-4-yloxy)-3-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]-2-propanol
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)propan-2-ol
SMILES [2H]C([2H])([2H])C([2H])(NCC(O)COc1cccc2[nH]c3ccccc3c12)C([2H])([2H])[2H]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H22N2O2/c1-12(2)19-10-13(21)11-22-17-9-5-8-16-18(17)14-6-3-4-7-15(14)20-16/h3-9,12-13,19-21H,10-11H2,1-2H3/i1D3,2D3,12D
Formula Weight 305.2121
Chemical Name or Material Carazolol D7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.