missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Carabin Polyclonal Antibody
GREENER_CHOICE

Product Code. 12818582
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818582 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818582 Supplier Invitrogen™ Supplier No. PA520394

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Daudi cell lysate. PA5-20394 can be used with blocking peptide PEP-0511.

Carabin is an endogenous inhibitor of calcineurin (see MIM 114105) that also inhibits the Ras (see MIM 190020) signaling pathway through its intrinsic Ras GTPase-activating protein activity (Pan et al., 2007 [PubMed 17230191]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carabin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TBC1D10C
Gene Accession No. Q8C9V1, Q8IV04
Gene Alias 1810062O14Rik; AI428527; CARABIN; EBP50-PDZ interactor of 64 kD; EPI64C; TBC1 domain family member 10C; TBC1 domain family, member 10c; TBC1D10C
Gene Symbols TBC1D10C
Host Species Rabbit
Immunogen A 16 amino acid peptide from near the carboxy terminus of human Carabin.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108995, 374403
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.