missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110996
This item is not returnable.
View return policy
Description
Immunogen sequence: EVHHDIREDV PPPKSRTSTA RSYIGSNHSS LGSMSPSNME GYSKTQYNQV PSEDFERTPQ SPTLPPAKVA APNLS Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 88%.
The protein encoded by this gene is a type I membrane receptor for group B coxsackieviruses and subgroup C adenoviruses. Pseudogenes of this gene are found on chromosomes 15, 18, and 21.
Specifications
| CAR | |
| Polyclonal | |
| Unconjugated | |
| CXADR | |
| 2610206D03Rik; 46 kD coxsackievirus and adenovirus receptor (CAR) protein; AU016810; AW553441; CAR; Car1; CAR4/6; coxsackie virus and adenovirus receptor; coxsackievirus and adenovirus receptor; Coxsackievirus and adenovirus receptor homolog; coxsackievirus B-adenovirus receptor; CVB3-binding protein; Cxadr; HCAR; HCVADR; MCAR; MCVADR; rCAR | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1525 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P78310 | |
| CXADR | |
| Recombinant Protein corresonding to Human CAR. Recombinant protein control fragment (Product #RP-95887). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction