missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAR/NR1I3 Antibody (OTI1C6), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71525IR
This item is not returnable.
View return policy
Description
CAR/NR1I3 Monoclonal specifically detects CAR/NR1I3 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| CAR/NR1I3 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NR1I3 | |
| Recombinant protein fragment corresponding to amino acids 1-236 of human CAR/NR1I3 (NP_001070950) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1C6 | |
| Western Blot, Immunohistochemistry | |
| CARCAR1, Constitutive activator of retinoid response, constitutive active receptor, Constitutive active response, Constitutive androstane receptor, MB67, MGC150433, MGC97144, MGC97209, nuclear receptor subfamily 1 group I member 3, nuclear receptor subfamily 1, group I, member 3, orphan nuclear hormone receptor, Orphan nuclear receptor MB67 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 9970.0 | |
| Store at 4°C in the dark. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido