missing translation for 'onlineSavingsMsg'
Learn More

Capzimin, MedChemExpress

Product Code. 16473056
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16473056

Brand: Medchem Express HY110404

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Capzimin is a potent and moderately specific proteasome isopeptidase Rpn11 inhibitor.

Capzimin is a potent and moderately specific proteasome isopeptidase Rpn11 inhibitor.

Specifications

Chemical Name or Material Capzimin
CAS 2084868-04-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H24N6O2S4
Quantity 25 mg
Solubility Information DMSO : ≥ 55.67 mg/mL (88.53 mM)
SMILES O=C(C1=CC2=CC=CC(SSC3=C4N=CC(C(NCCC5=NC=CS5)=O)=CC4=CC=C3)=C2N=C1)NCCC6=NC=CS6
Molecular Weight (g/mol) 628.81
Formula Weight 628.81
Percent Purity 99.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.