missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Capzimin dimer

Description
Capzimin dimer is a specific inhibitor of proteasome isopeptidase Rpn11 (IC50 = 2.1 μM and 3.8 μM, in 293T and A549 cell lines, respectively). It exhibits 80-fold selectivity for Rpn11 over Csn5, ten-fold over AMSH and six-fold over BRCC36 (IC50 = 30 μM, 4.5 μM, and 2.3 μM, respectively). Capzimin dimer blocks proliferation of cancer cells.
Specifications
Specifications
| Chemical Name or Material | 8,8 '-Dithiobis[N-[2-(2-thiazolyl)ethyl]-3-quinolinecarboxamide] |
| CAS | 2084868-04-0 |
| Quantity | 10 mg |
| Target | Proteasome Inhibitors |
| Molecular Formula | C30H24N6O2S4 |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?