missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPZA3 Polyclonal Antibody

CAPZA3 Polyclonal Antibody
Brand: Invitrogen PA570405
This item is not returnable.
View return policy
Description
CAPZA3 Polyclonal Antibody for Western Blot
Specifications
| CAPZA3 | |
| Polyclonal | |
| Unconjugated | |
| CAPZA3 | |
| 510-4; CAPAA3; Cappa3; capping actin protein of muscle Z-line alpha subunit 3; capping actin protein of muscle Z-line subunit alpha 3; capping protein (actin filament) muscle Z-line, alpha 3; capping protein alpha 3; capZ alpha-3; Capza3; cp alpha3; CP-alpha-3; epididymis secretory protein Li 86; F-actin capping protein alpha-3 subunit; F-actin-capping protein subunit alpha-3; Germ cell-specific protein 3; GSG3; HEL-S-86; repro32; testis tissue sperm-binding protein Li 73n; Tex8 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 93661 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96KX2 | |
| CAPZA3 | |
| synthetic peptide directed towards the N terminal region of human CAPZA3. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction