missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAPZA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117832
This item is not returnable.
View return policy
Description
CAPZA2 Polyclonal Antibody for Western Blot, IP
The protein encoded by this gene is a member of the F-actin capping protein alpha subunit family. It is the alpha subunit of the barbed-end actin binding protein Cap Z. By capping the barbed end of actin filaments, Cap Z regulates the growth of the actin filaments at the barbed end.
Specifications
| CAPZA2 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P47755 | |
| Capza2 | |
| E. coli-derived Human CAPZA2 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Capza2 | |
| 1110053K06Rik; AW208808; Cappa2; capping actin protein of muscle Z-line alpha subunit 2; capping protein (actin filament) muscle Z-line, alpha 2; capping protein alpha 2; CAPZ; capZ alpha-2; CAPZA2; CAZA2; F-actin capping protein alpha-2 subunit; F-actin-capping protein subunit alpha-2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 830 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction