missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPZA1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55145-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to CAPZA1.
CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.Specifications
| CAPZA1 | |
| Polyclonal | |
| Unconjugated | |
| CAPZA1 | |
| CAPPA1, CAPZ, CapZ alpha 1, CAPZA1, CAZ1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12340, 829 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P47753, P52907 | |
| CAPZA1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction