missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAPZA1 Monoclonal Antibody (A2E7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA536109
This item is not returnable.
View return policy
Description
CAPZA1 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot
CAPZA1 is a member of the F-actin capping protein alpha subunit family. This gene encodes the alpha subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
Specifications
| CAPZA1 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P52907 | |
| CAPZA1 | |
| Recombinant protein within Human CAPZA1 aa 60-248. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| A2E7 | |
| Unconjugated | |
| CAPZA1 | |
| Cap Z; Cappa1; capping actin protein of muscle Z-line alpha subunit 1; capping actin protein of muscle Z-line subunit alpha 1; capping protein (actin filament) muscle Z-line, alpha 1; capping protein alpha 1; CAPZ; CapZ alpha-1; capZ alpha-1 {ECO:0000250; CAPZA1; capza1 {ECO:0000250; CAZ1; F-actin capping protein alpha-1 subunit; F-actin-capping protein subunit alpha-1; F-actin-capping protein subunit alpha-1 {ECO:0000250; UniProtKB:P52907}; unnamed protein product | |
| Mouse | |
| Protein G | |
| RUO | |
| 829 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction