missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAPZA1 Antibody (2C8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42937
This item is not returnable.
View return policy
Description
CAPZA1 Monoclonal antibody specifically detects CAPZA1 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| CAPZA1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 2C8 | |
| Western Blot 1:1000 - 1:8000, ELISA, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| Cap Z, CAPPA1, capping protein (actin filament) muscle Z-line, alpha 1, CAPZ, CapZ alpha-1, CAZ1, F-actin-capping protein subunit alpha-1 | |
| capping protein(actin filament) muscle Z-line, alpha 1 - (Uniprot# P52907) | |
| 100 μg | |
| Extracellular Matrix | |
| 829 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction