missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ IgG-Fc (Multispecies) Magnetic Agarose Beads

Product Code. 18486598 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18486598 1 mL 1mL
18436458 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18486598 Supplier Thermo Scientific™ Supplier No. 2882852001
  • ONLINE EXCLUSIVE PRICE
    £450.00
    £428.00 / 1mL
    Save £22.00
    4% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ IgG-Fc (Multispecies) Magnetic Agarose Beads are high-capacity, high-throughput magnetic affinity particles for antibody purification using manual or robotic magnetic separators.

CaptureSelect IgG-Fc (Multispecies) Magnetic Agarose Beads are high-capacity, high-throughput magnetic affinity particles for antibody purification using manual or robotic magnetic separators. The CaptureSelect IgG-Fc (multispecies) ligand has been specifically designed to purify IgG through high-affinity binding of the IgG Fc region. These magnetic agarose beads can be used for the purification of IgG from a broad range of species. The tested species and their specificity are shown in the figure below.

Features of CaptureSelect IgG-Fc (multispecies) Magnetic Agarose Beads include:

  • One-step IgG purification with high purity, from crude material
  • Compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher instruments)
  • Short operation time
  • No sample dilution required during isolation, repetitive sample loading feasible
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: 6% cross-linked magnetized agarose
  • Average particle size: 25–30 μm
  • Suspension: 25% slurry
  • Ligand: CaptureSelect IgG-Fc (multispecies) affinity ligand
  • Binding capacity: >4 mg IgG/mL slurry
  • Elution conditions: 0.1 M glycine, pH 3.0

Specifications

Formulation 25% (v/v) slurry in Tris buffered saline with 0.05% Tween 20 and 0.05% sodium azide
Matrix 6% cross-linked magnetized agarose
Content And Storage Store at 2°C to 8°C.
Shipping Condition Wet Ice
Particle Size 25 to 30 μm
Form Resin Slurry
Product Line CaptureSelect
Quantity 1 mL
Type Magnetic Affinity Bead
Ligand Type CaptureSelect IgG-Fc (multispecies) Affinity Ligand
Sufficient For Sample Preparation
Binding Capacity >4 mg IgG/mL Slurry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.