missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ CH1-XL MiniChrom™ Pre-packed Column

Product Code. 18416478 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
18416478 1 mL 1mL
18446428 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18416478 Supplier Thermo Scientific™ Supplier No. 5943462001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ CH1-XL MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

CaptureSelect CH1-XL MiniChrom™ columns are designed for bench-scale resin screening, process development, and sample preparation.

These pre-packed columns are ready to connect and have the following features:

  • Dimensions of 1 mL (0.8 x 2 cm) and 5 mL (0.8 x 10 cm)
  • Direct connection to standard chromatography systems
  • Packed with CaptureSelect CH1-XL Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates

The 1 mL columns can be used for quick screenings of application feasibility and lab scale purification on a convenient and easy-to-use pre-packed column. The 10 cm bed height of the 5 mL columns allows initial process development on a bench scale.

The CaptureSelect CH1-XL Affinity Matrix recognizes the CH1 domain of human IgG antibodies, thereby enabling purification of all Fab fragments, independent of the light-chain type (kappa or lambda) and without binding of over-expressed light chain and light-chain dimers. In addition, it can be used for the purification of human IgGs from all four subclasses (1, 2, 3, and 4).

Features of this affinity matrix include:

  • Platform approach for purification of high quality and correctly assembled human Fab fragments
  • No co-purification of over-expressed free light chains due to selectivity for the CH1 domain
  • Elution possible at pH 4.0–4.5 with 50 mM acetate buffer
  • Caustic cleaning with up to 50 mM NaOH
  • Excellent scalability
  • No cross binding to bovine IgG, therefore presence of fetal bovine serum is not a problem

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect CH1-XL affinity ligand
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >15 mg/mL human Fab fragment, depending on flow rate, column height, and contact time. Note that humanized Fab fragments may have lower capacities.
  • Elution conditions: 100 mM glycine buffer, pH 3.0; 50 mM acetate buffer, pH 4.0 - 4.5
  • Column dimensions: 1 mL (0.8 x 2 cm) and 5 mL (0.8 x 10 cm)
  • Flow characteristics: 75–150 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol

Specifications

Column Format Chromatography Cartridge
Diameter (Metric) 0.8 cm
Flow Rate 75 to 150 cm/hr. (Up to 2 bar)
Length (Metric) 2 cm
Ligand Type CH1-XL affinity ligand
Particle Size 65 ± 10 μm
Quantity 1 mL
Stationary Phase CaptureSelect CH1-XL affinity ligand
Content And Storage Store in refrigerator (2°C to 8°C).
Product Line CaptureSelect
Binding Capacity >15 mg/mL human Fab fragment
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Matrix Agarose-based, epoxide-activated
Volume (Metric) 1 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.