Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-IgG4 (Human) Conjugate
Alle ansehen Thermo Scientific ProdukteBeschreibung
CaptureSelect Biotin Anti-IgG4 (Hu) Conjugate consists of a 12 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds to the Fc part of the human IgG4 subclass. The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces.
The CaptureSelect Biotin Anti-IgG4 (Hu) Conjugate allows you to:
- Detect, quantitate, and characterize human IgG4 antibodies
- Avoid cross-binding with other IgG subclasses like IgG1, 2 and 3 and isotypes like IgM and IgA including IgG from bovine sources such as FCS
Applications for CaptureSelect Biotin Anti-IgG4 (Hu) Conjugate include ELISA, Western blot, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Spezifikation
Spezifikation
| Product Type | Biotin Anti-IgG4 Conjugate |
| Conjugate | Biotin and Other Haptens |
| Quantity | 100 μg |
| Product Line | CaptureSelect |
| Form | Liquid |
| Content And Storage | Store in freezer (-5°C to -30°C). |
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.