missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ AAV8 Ligand Leakage ELISA

Product Code. p-7180722 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 Assay
10 Assays
Unit Size:
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 18466638

Brand: Thermo Scientific™ 810338001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ CaptureSelect™ AAV8 Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAV8 Affinity Resin.

CaptureSelect AAV8 Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAV8 Affinity Resin. In this resin the ligand, a 13 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the purified AAV.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect AAV8 Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications

Content And Storage 3 x 0.1 mL vials, store at -5°C to -30°C
Detection Method Colorimetric
Product Line CaptureSelect
For Use With (Equipment) Microplate Reader
Quantity 1 Assay
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.