Learn More
Captopril, Thermo Scientific™
Orally active inhibitor of angiotensin-converting enzyme
£43.50 - £425.00
Chemical Identifiers
| CAS | 62571-86-2 |
|---|---|
| Molecular Formula | C9H15NO3S |
| Molecular Weight (g/mol) | 217.283 |
| MDL Number | MFCD00168073 |
| InChI Key | FAKRSMQSSFJEIM-RQJHMYQMSA-N |
| Synonym | captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril |
| PubChem CID | 44093 |
| ChEBI | CHEBI:3380 |
| IUPAC Name | (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid |
| SMILES | CC(CS)C(=O)N1CCCC1C(=O)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15435789
|
Thermo Scientific Alfa Aesar
J63593.03 |
1g |
£43.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15445789
|
Thermo Scientific Alfa Aesar
J63593.06 |
5g |
£132.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15455789
|
Thermo Scientific Alfa Aesar
J63593.14 |
25g |
£425.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsCaptopril has also been shown to inhibit the formation of angiotensin II, a bioactive peptide that stimulates angiogenesis and increases microvessel density. Captopril demonstrates noncompetitive inhibition of tyrosinase monophenolase activity and competitive inhibition of diphenolase activity
Solubility
Soluble in water (160 mg/ml) at 25°C, sesame and corn oils (<1mg/mL) at 25°C, methanol (>100 mg/ml), ethanol (>100 mg/ml), isopropanol (>100 mg/ml), chloroform (>100 mg/ml), and dichloromethane (>100 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
| 62571-86-2 | |
| 217.283 | |
| FAKRSMQSSFJEIM-RQJHMYQMSA-N | |
| 44093 | |
| (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid |
| C9H15NO3S | |
| MFCD00168073 | |
| captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril | |
| CHEBI:3380 | |
| CC(CS)C(=O)N1CCCC1C(=O)O |
Specifications
| 62571-86-2 | |
| MFCD00168073 | |
| 14,1774 | |
| Soluble in water (160mg/ml) at 25°C,sesame and corn oils (<1mg/ml) at 25°C,methanol (>100mg/ml),ethanol (>100mg/ml),isopropanol (>100mg/ml),chloroform (>100mg/ml),and dichloromethane (>100mg/ml). | |
| CC(CS)C(=O)N1CCCC1C(=O)O | |
| 217.283 | |
| CHEBI:3380 | |
| Captopril |
| C9H15NO3S | |
| 477887 | |
| captopril, capoten, l-captopril, captopryl, lopirin, cesplon, captolane, tensoprel, acepress, captoril | |
| FAKRSMQSSFJEIM-RQJHMYQMSA-N | |
| (2S)-1-[(2S)-2-methyl-3-sulfanylpropanoyl]pyrrolidine-2-carboxylic acid | |
| 44093 | |
| 217.29 |
Safety and Handling
GHS H Statement
H361-H317
Suspected of damaging fertility or the unborn child.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P305+P351+P338-P308+P313-P310-P333+P313-P363-P501c
H317-H318-H360D
missing translation for 'einecsNumber' : 263-607-1
missing translation for 'rtecsNumber' : UY0550000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.