missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ Multimodal (MMC) Chromatography Media

Product Code. 10089269 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10089269 100 mL 100mL
10681521 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10089269 Supplier Cytiva Supplier No. 17531702
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Used to solve specific purification challenges at both high and low ionic strengths

  • Composed of rigid, high-flow agarose matrix and novel multimodal ligand
  • Used to bind proteins at conductivity of feed material
  • Ligand contains carboxylic acid group and exhibit the properties of weak cation exchanger
  • High dynamic binding capacity at high conductivity
  • High-volume throughput cuts process times
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
  • Dynamic binding capacity at 10% breakthrough as measured at a residence time of 2 min, 300cm/h in a Tricorn(TM) 5/100 column with 10cm bed height in 50mM Sodium acetate, pH 4.75, 250mM NaCl.

Warranty

Store at 4° to 30°C in 20% Ethanol

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Stability 1 M Acetic Acid, 1 M Sodium Hydroxide, 8 M Urea, 6 M Guanidine Hydrochloride, and 70% Ethanol
For Use With (Application) Used to Solve Specific PurifiCatIon Challenges at Both High and Low Ionic Strengths
Storage Requirements 4°C to 30°C
Binding Capacity >45 mg BSA/mL medium (at 30 mS/cm2)
Product Type Capto MMC Media
Flow Rate 600 cm/Hoμr
Particle Size 75 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Ionic Capacity 0.07 to 0.09 mM H+/mL
Ion Exchange Type Multimodal Weak CatIon Exchanger
Max. Pressure 300 kPa
Matrix Highly cross-linked agarose
pH Range 2 to 14
Storage Buffer 20% Ethanol
Ligand Type Carboxylic Acid Group
Show More Show Less
compliance-icons
Product Identifier
  • Capto MMC, 100 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.