missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- High yields achieved through high-resolution beads and ligand selectivity
- Efficient removal of aggregates, viruses, and main contaminants in MAb processes
- Enables use of platform approach to MAb process development
- Displays broad pH/salt operational window
Avoid using oxidizing agents, cationic detergents.
Specifications
Specifications
| Quantity | 25 mL |
| Stability | 1 M Acetic Acid, 1 M NaOH, 2 M NaCl, 5% 1-Propanol, 30% Isopropanol, 70% Ethanol |
| For Use With (Application) | For High Resolution Polishing of Monoclonal Antibodies (MAbs) and Other Biomolecules |
| Storage Requirements | 4°C to 30°C |
| Anion Exchange Capacity | 25 to 39 μM/mL |
| Binding Capacity | 60 to 90 mg Mab/mL |
| Product Type | Capto MMC ImpRes Media |
| Particle Size | 40 μm |
| Max. Pressure | 300 kPa |
| Matrix | High-flow Agarose |
| Show More |
Safety and Handling
Product Identifier
- Capto MMC ImpRes, 25 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?