missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Capto™ DEAE Chromatography Media

Product Code. 10019389 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10019389 100mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10019389 Supplier Cytiva Supplier No. 17544301
Outlet Corner
Use promo code 28080
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Used for fast, efficient capture and intermediate protein purification

  • Composed of high-flow agarose matrix modified with dextran surface extenders and weak diethylaminoethyl (DEAE) anion exchanger
  • Combination of high volume throughput and dynamic binding capacity at high flow
  • Withstands effective CIP procedure and is chemically stable
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

TRUSTED_SUSTAINABILITY

Specifications

Quantity 100mL
For Use With (Application) For Capture, Intermediate Protein Purification
Anion Exchange Type Weak anion exchanger
Product Type Capto DEAE Media
Particle Size 90 μm
Matrix Cross linked agarose w/dextran surface extender
compliance-icons
Product Identifier
  • Capto DEAE, 100 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.