Learn More
Cytiva Capto™ adhere ImpRes Multimodal BioProcess™ Chromatography Medium
For high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules
Brand: Cytiva 17371501
Code : GE
Additional Details : Weight : 0.07600kg
Includes: Certificate of Analysis
Description
The strong anion exchange multimodal ligand displays high selectivity compared with traditional ion exchanger, which allows the possibility to solve challenging purification problems. Main contaminants in MAb processes such as DNA, host cell proteins (HCP), leached protein A, aggregates, and viruses are efficiently separated.
- High-resolution beads and ligand selectivity ensures high yield
- Efficient removal of aggregates, viruses, and main contaminants in MAb processes
- Enables use of platform approach to MAb process development
- Allows separation of MAb charged variants
Avoid using oxidizing agents, anionic detergents.
Specifications
25 mL | |
1 M Acetic Acid, 1 M NaOH, 2 M NaCl, 5% 1-Propanol, 30% Isopropanol, 70% Ethanol | |
0.08 to 0.11 mM Cl-/mL | |
Capto Adhere ImpRes Media | |
Certificate of Analysis | |
High-flow Agarose | |
20% Ethanol |
For High Resolution Polishing of Monoclonal Antibodies (MAbs) and Other Biomolecules | |
4°C to 30°C | |
45 to 85 mg Mab/mL | |
40 μm | |
300 kPa | |
3 to 12 | |
Multimodal Strong AnIon Exchanger |