Learn More
Erlab™ Captair™ Work Surfaces
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ Work Surfaces are for use with Captair PCR work stations.
Brand:  Erlab™ I0117030001
Code : 50
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12878733
  • £1022.90 / Each
Estimated Shipment: 19-07-2024
to see stock.
Description
Description
  • High chemical and mechanical resistance
  • Rounded corners to facilitate cleaning operations
  • Built in spill tray
Specifications
Specifications
For Captair™ Flex™ Model XLS392 fume hood
Built-in Retention Tray
Work surface No. 2
Phenolic Resin