Learn More
Erlab™ Captair Smart Glass Work Surface with Retention Tray
Erlab™ Captair Smart Work Surface with Retention Tray is designed to be used with the range of Captair Smart Ductless Filtering Fume Hoods.
Brand:  Erlab™ I0165020001
Additional Details : Weight : 25.00000kg
 View more versions of this product
Product Code. 15555164
  • £969.90 / Each
Estimated Shipment: 09-08-2024
to see stock.
Description
Description
  • Tempered enameled glass worktop
  • Includes retention tray filtering fume hoods
  • Jointed frame
  • Ergonomic arm rest for a comfortable working position
Specifications
Specifications
Smart 633 Fume Hood
585 mm
1503 mm
Tempered Glass
Retention Tray
Jointed Frame
Work Surface