Learn More
Erlab™ Captair by Erlab™ H402 Version 03 Filtration Columns
Product Code. 12088568
Click to view available options
Filter Type:
BE Filter
F Carbon Filter
Unit Size:
Each
Product Code. 12088568
Brand: Erlab™ G0103/F
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for laboratories specializing in fields such as biology, botany, aerospace, electronics, pharmaceutical and cosmetics. Erlab™ Captair™ H402 Version 03 Filtration Columns are for storing organic vapors and acids
  • Horizontal filtration column
  • HEPA H14 filter
  • Handlings in an ultra clean environment
  • Serial prefilter(optimizes the HEPA H14 filter lifetime)
  • Optional carbon filtration = clean the air polluted by ambient air VOCs
Specifications
Product Type Filtration Column
Filter Type F Carbon Filter
For Use With (Application) For storing Formaldehyde