missing translation for 'onlineSavingsMsg'
Learn More

Erlab Captair 483 Smart PreFilters, Pack of 3

Product Code. 16615155
Change view
Click to view available options
For Use With (Equipment):
Storage Cabinet
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
16615155 Storage Cabinet Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16615155 Supplier Erlab Supplier No. I1064000001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prefilter for your Ductless fume hoods.

  • Prefilter for your Ductless fume hood Captair 483 Smart
  • Dimensions: 336 x 362 x 10 mm

Specifications

For Use With (Equipment) Storage Cabinet
Product Type Prefilter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.