missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erlab Filtering Storage Cabinet Captair Smart
Click to view available options
Model:
1634
632B
834
Construction:
Pull-out Doors
Sliding Doors
Swing Doors
Unit Size:
Each
Description
This filtering chemical storage cabinets Captair Smart provides outstanding filtration quality. We ensure users and laboratory technicians are protected against chemical hazards.
- Safety Standards: Filtration performances tested according to the AFNOR NF X 15 211:2009 standard : France - EN 1822 : 1998 (HEPA H14 and ULPA U17 Filters)
- CE Marking
- Dimensions (mm): L 900 x D 660 x H 2140
- Air Flow 220 m3/h / 129 CFM
- Voltage / Frequency 110-230 V / 50-60 Hz
- Power consumption 45 W
- Structure: Corrosion resistant electro-galvanized steel coated with anti-acid polymer
- Doors: Clear, chemical resistant acrylic for easy viewing
- Filtration module is made with injected polypropylene
- Filtration technology 1 column that can be configured to handle liquids, powders, or both
- Following filtration column configuration: Carbon filtration for gases and vapors and particulate filtration for powders
- Detection sensor for : Type S, for solvents / Type A, for acids / Type F, for formaldehydes
- Temperature/Relative Humidity sensor
- Capteur
- Door sensors: Alarm if doors are left open
- Storage compartments: 2
- Delivered with 10 adjustable shelves with integrated retention tray
- Absorbing mats: 2
- Storage capacities: 100 x 1L glass bottles
- Key lock
- Communication Interface: When the light is pulsing: Doors left open - Containment is compromised - Filter breakthrough
- eGuard APP: For remote control to monitor the status of the storage cabinet, change the settings, and deliver safety alerts immediately to your devices
- Network connection
Specifications
Specifications
| Product Type | Ductless Filtering Fume Hood |
| Filter Type | Bonded Carbon and/or HEPA |
| Material | Corrosion Resistant Electro-galvanized Steel, Antiacid Polymer Coating |
| Construction | Swing Doors |
| Model | 1634 |
| Voltage | 110/230 V |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction