missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAPSL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569866
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
CAPSL gene ontology annotations related to this gene include calcium ion binding.
Specifications
| CAPSL | |
| Polyclonal | |
| Unconjugated | |
| Capsl | |
| 1700028N11Rik; AW125205; Calcyphosine; calcyphosine like; calcyphosine-like; calcyphosin-like protein; CAPSL; RGD1308776 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 294795 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| Capsl | |
| synthetic peptide corresponding to a region of rat CAPSL. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction