missing translation for 'onlineSavingsMsg'
Learn More

Capsazepine, MedChemExpress

Product Code. 16472787
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472787 50 mg 50mg
16466856 100 mg 100mg
16456708 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16472787 Supplier Medchem Express Supplier No. HY15640
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Capsazepine is a synthetic analogue of the sensory neurone excitotoxin, and an antagonist of TRPV1 receptor with an IC50 of 562 nM.

Capsazepine is a synthetic analogue of the sensory neurone excitotoxin, and an antagonist of TRPV1 receptor with an IC50 of 562 nM.

Specifications

Chemical Name or Material Capsazepine
CAS 138977-28-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H21ClN2O2S
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (265.32 mM)
SMILES S=C(N1CCCC2=CC(O)=C(O)C=C2C1)NCCC3=CC=C(Cl)C=C3
Molecular Weight (g/mol) 376.9
Formula Weight 376.9
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.