missing translation for 'onlineSavingsMsg'
Learn More

Capsazepine, 98+%, Thermo Scientific™

A competitive antagonist for capsaicin and resiniferatoxin

£185.00 - £711.00

Chemical Identifiers

CAS 138977-28-3
Molecular Formula C19H21ClN2O2S
Molecular Weight (g/mol) 376.899
MDL Number MFCD00153778
InChI Key DRCMAZOSEIMCHM-UHFFFAOYSA-N
Synonym capsazepine, capsazepin, n-2-4-chlorophenyl ethyl-1,3,4,5-tetrahydro-7,8-dihydroxy-2h-2-benzazepine-2-carbothioamide, unii-lfw48my844, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide, 2-2-4-chlorophenyl ethylamino-thiocarbonyl-7,8-dihydroxy-2,3,4,5-tetrahydro-1h-2-benzazepine, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2h-2-benzazepine-2-carbothioamide, capasazepine, tocris-0464
PubChem CID 2733484
ChEBI CHEBI:70773
IUPAC Name N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide
SMILES C1CC2=CC(=C(C=C2CN(C1)C(=S)NCCC3=CC=C(C=C3)Cl)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15405329
SDS View Documents
Thermo Scientific Alfa Aesar
J63055.MA
10mg
ONLINE EXCLUSIVE PRICE
£189.00
£185.00 / 10mg
Save £4.00
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15415329
SDS View Documents
Thermo Scientific Alfa Aesar
J63055.MF
50mg
ONLINE EXCLUSIVE PRICE
£727.00
£711.00 / 50mg
Save £16.00
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Capsazepine is shown to inhibit the development of mechanical hyperalgesia induced by intraplanar capsaicin injection. Capsazepine is described to block Resiniferatoxin (sc-24015)- and Capsaicin-induced contractions of guinea pig treacheal smooth muscle. Studies investigating the function of the VR1 (TRPV1 receptor) have employed Capsazepine as a tool for perturbing systemic neuronal response to Capsaicin.

Solubility
Soluble in ethanol or DMSO

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

138977-28-3
376.899
DRCMAZOSEIMCHM-UHFFFAOYSA-N
2733484
N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide
C19H21ClN2O2S
MFCD00153778
capsazepine, capsazepin, n-2-4-chlorophenyl ethyl-1,3,4,5-tetrahydro-7,8-dihydroxy-2h-2-benzazepine-2-carbothioamide, unii-lfw48my844, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide, 2-2-4-chlorophenyl ethylamino-thiocarbonyl-7,8-dihydroxy-2,3,4,5-tetrahydro-1h-2-benzazepine, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2h-2-benzazepine-2-carbothioamide, capasazepine, tocris-0464
CHEBI:70773
C1CC2=CC(=C(C=C2CN(C1)C(=S)NCCC3=CC=C(C=C3)Cl)O)O

Specifications

138977-28-3
MFCD00153778
Soluble in ethanol or DMSO
C1CC2=CC(=C(C=C2CN(C1)C(=S)NCCC3=CC=C(C=C3)Cl)O)O
376.899
CHEBI:70773
≥99%
C19H21ClN2O2S
capsazepine, capsazepin, n-2-4-chlorophenyl ethyl-1,3,4,5-tetrahydro-7,8-dihydroxy-2h-2-benzazepine-2-carbothioamide, unii-lfw48my844, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide, 2-2-4-chlorophenyl ethylamino-thiocarbonyl-7,8-dihydroxy-2,3,4,5-tetrahydro-1h-2-benzazepine, n-2-4-chlorophenyl ethyl-7,8-dihydroxy-1,3,4,5-tetrahydro-2h-2-benzazepine-2-carbothioamide, capasazepine, tocris-0464
DRCMAZOSEIMCHM-UHFFFAOYSA-N
N-[2-(4-chlorophenyl)ethyl]-7,8-dihydroxy-1,3,4,5-tetrahydro-2-benzazepine-2-carbothioamide
2733484
376.9
Capsazepine
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.