missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capsaicin, MedChemExpress
Description
Capsaicin ((E)-Capsaicin), an active component of chili peppers, is a TRPV1 agonist. Capsaicin has pain relief, antioxidant, anti-inflammatory, neuroprotection and anti-cancer effects.
Specifications
Specifications
| Chemical Name or Material | Capsaicin |
| CAS | 404-86-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H27NO3 |
| Quantity | 100 mg |
| Synonym | (E)-Capsaicin |
| Solubility Information | Ethanol : 200 mg/mL (654.86 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (327.43 mM; Need ultrasonic) |
| SMILES | CC(C)/C=C/CCCCC(NCC1=CC=C(O)C(OC)=C1)=O |
| Molecular Weight (g/mol) | 305.41 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?