missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Capsaicin 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Capsaicin |
| CAS | 404-86-4 |
| Analyte or Component Names | Capsaicin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H27 N O3 |
| Concentration | 10 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Capsaicin, 6-Nonenamide, 8-methyl-N-vanillyl-, (E)- (8CI), 6-Nonenamide, N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methyl-, (E)-, Capsaicin (6CI), (6E)-N-[(4-Hydroxy-3-methoxyphenyl)methyl]-8-methyl-6-nonenamide, (E)-8-Methyl-N-vanillyl-6-nonenamide, (E)-N-(4-Hydroxy-3-methoxybenzyl)-8-methylnon-6-enamide, ALGRX 4975, Adlea, Ausanil, Axsain, Capsaicine, Capsicin, Capsin P 50, Capzasin-HP, Chilli Orange, Dolenon, E-Capsaicin, Mioton, N-(4-Oxy-3-methoxybenzyl)-8-methyl-6-nonenamide, NGX 4010, NSC 56353, Ovocap, Qutenza, Ratden PE 40, Togarashi Orenji, Zostrix, trans-8-Methyl-N-vanillyl-6-nonenamide, trans-Capsaicin |
| InChI Formula | InChI=1S/C18H27NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h6,8,10-12,14,20H,4-5,7,9,13H2,1-3H3,(H,19,21)/b8-6+ |
| SMILES | C(NC(CCCC/C=C/C(C)C)=O)C1=CC(OC)=C(O)C=C1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction