Learn More
Invitrogen™ CAPS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA5140064
Descrizione
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LGSLVDSDDE DNFSYIPLST ANLPNSSSTL GWVTPCQTPY TQYHLNKLDQ NIIPENLPAP TDKCKLKYQQ CKTEIKEGYK QYSQRNAENT KSNVT Highest antigen sequence indentity to the following orthologs: Mouse - 54%, Rat - 57%.
Calcyphosine-2 is a calcium-binding protein with 2 EF-hand motifs (Wang et al., 2002 [PubMed 11846421]).
Specifica
| CAPS2 | |
| Polyclonal | |
| Unconjugated | |
| CAPS2 | |
| calcyphosin 2; calcyphosin-2; calcyphosine 2; calcyphosine-2; calcyphosphine 2; CAPS2; D630005B03Rik; UG0636c06 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84698 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXY5 | |
| CAPS2 | |
| Recombinant protein corresponding to Human CAPS2. Recombinant protein control fragment (Product #RP-108824). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.