Learn More
Invitrogen™ CAPS1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA568381
Product Code. 15813224
  • £449.00 / 100µL
Description
CAPS1 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a novel neural/endocrine-specific cytosolic and peripheral membrane protein required for the Ca2+-regulated exocytosis of secretory vesicles. The protein acts at a stage in exocytosis that follows ATP-dependent priming, which involves the essential synthesis of phosphatidylinositol 4,5-bisphosphate P2). Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CAPS1
Polyclonal
Unconjugated
CADPS
AU067781; Ca++-dependent secretion activator; Ca>2+>dependent activator protein for secretion; Ca>2+-dependent activator protein for secretion; Ca2+ dependent secretion activator; Ca2+-dependent activator protein; Ca2+-dependent activator protein for secretion; Ca2+-dependent secretion activator; Ca2+-regulated cytoskeletal protein; Cadps; CADPS1; calcium dependent secretion activator; calcium-dependent activator protein for secretion 1; Calcium-dependent secretion activator 1; Caps; Caps1; CAPS-1; Caps2; KIAA1121; mKIAA1121; rCAPS; UNC-31
Rabbit
Antigen affinity chromatography
RUO
8618
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9ULU8
CADPS
Synthetic peptide corresponding to residues near the N terminal of human CAPS.