missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Caps for Transferpettor™

Product Code. 10490413 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
5 μL
20 μL
25 μL
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10490413 20 μL Pack of 100
10343631 5 μL Pack of 100
10430993 25 μL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10490413 Supplier BRAND™ Supplier No. 701904
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

DE-M certified. Includes color code. Glass construction.

  • For use with Transferpettor™
  • DE-M marking
  • Glass construction
  • Includes color code
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DE-M certified
Color Black
Material Glass
Volume (Metric) 20 μL
Quantity 100 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.