missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Caps for Sample Cups

Product Code. 31922524
Change view
Click to view available options
Closure Type:
Flat Top
Pierceable Cross-cut
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type Unit Size
31922524 Flat Top Pack of 1000
31922388 Pierceable Cross-cut Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31922524 Supplier Globe Scientific Supplier No. 113132
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Designed specifically for use on Globe Scientific multi-purpose sample cups. Globe Scientific Caps for Sample Cups are made from polyethylene and snap securely into place. Caps are available with a solid or cross-cut pierceable top.

  • For use on Globe Scientific sample cups
  • Snap easily and securely into place
  • Available with flat top (21-200-100) or pierceable cross cut (21-200-101)
  • Polyethylene (PE)
  • Natural color

Specifications

Closure Type Flat Top
Quantity 1000/Pk.
Color Natural
Product Type Snap Cap
Material Polyethylene
For Use With (Equipment) Globe Scientific Sample Cups
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.