missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CAPS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118991
This item is not returnable.
View return policy
Description
CAPS Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| CAPS | |
| Polyclonal | |
| Unconjugated | |
| CAPS | |
| Calcyphosin; calcyphosine; calcyphosine 1; CAPS; CAPS1; thyroid protein p24 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100514564, 828 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q13938 | |
| CAPS | |
| Recombinant protein Calcyphosine. The antigen corresponds to amino acid range 1-189 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction