missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford CAPS
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
pKa (25°C): 10.4. The working pH range in aqueous solution is 9.7-11.1
Specifications
Specifications
| Chemical Name or Material | CAPS |
| CAS | 1135-40-6 |
| Quantity | 100 g |
| Molecular Formula | C9H19NO3S |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction