missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caprophenone, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Caprophenone |
| CAS | 942-92-7 |
| Molecular Formula | C12H16O |
| Quantity | 25 g |
| Synonym | Hexanophenone, 1-Phenyl-1-hexanone, 1-Benzoylpentane, NSC 8480, Pentyl Phenyl Ketone, Phenyl Pentyl Ketone, n-Pentyl Phenyl Ketone |
| InChI Formula | InChI=1S/C12H16O/c1-2-3-5-10-12(13)11-8-6-4-7-9-11/h4,6-9H,2-3,5,10H2,1H3 |
| SMILES | CCCCCC(=O)c1ccccc1 |
| IUPAC Name | 1-phenylhexan-1-one |
| Molecular Weight (g/mol) | 176.25 |
| Formula Weight | 176.1201 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction