Learn More
MP Biomedicals™ Capreomycin Sulfate, MP Biomedicals™
Often grouped with aminoglycosides. Capreomycin Sulfate, MP Biomedicals is a cyclic peptide antibiotic. This second line antibiotic binds across the ribosomal interface involving 23S rRNA helix 69 (H69) and 16S rRNA helix 44 (h44).
Brand: MP Biomedicals™ 0215492491
Code : NEW
Additional Details : CAS Number : 1405-37-4 Weight : 0.00100kg
Description
Capreomycin is used to study multidrug-resistant tuberculosis. It is also used to study bacterial ribosomal subunit interactions and translocation processes during protein synthesis.
Specifications
1405-37-4 | |
Powder | |
750.79 | |
Streptomyces capreolus |
-20°C | |
Capreomycin is a second line antibiotic and is used to study multidrug-resistant tuberculosis. Capreomycin sulfate is used to study bacterial ribosomal subunit interactions and translocation processes during protein synthesis. | |
1 g |
Safety and Handling
- Capreomycin Sulfate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Capreomycin Sulfate