missing translation for 'onlineSavingsMsg'
Learn More

Capreomycin sulfate, MedChemExpress

Product Code. 16451699
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16451699

Brand: Medchem Express HY17566

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Capreomycin sulfate is a peptide antibiotic, commonly grouped with the aminoglycosides, which is given in combination with other antibiotics for MDR-tuberculosis.

Capreomycin sulfate is a peptide antibiotic, commonly grouped with the aminoglycosides, which is given in combination with other antibiotics for MDR-tuberculosis.

Specifications

Chemical Name or Material Capreomycin sulfate
CAS 1405-37-4
Color White
Purity Grade Notes Research
Molecular Formula C25H46N14O11S
Quantity 1 g
Solubility Information H2O : ≥ 37 mg/mL (49.28 mM)
SMILES O=C(CC(N)CCCN)NCC(C(N/C(C(NC(C1NC(NCC1)=N)C2=O)=O)=C/NC(N)=O)=O)NC(C(NC(C(CN2)N)=O)C)=O.O=S(O)(O)=O
Molecular Weight (g/mol) 750.78
Formula Weight 750.78
Percent Purity 98.07%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.